Microscope Slides, Ground 90°
Product Image
Fisherbrand
Microscope Slides, Ground 90°
Premium slides are manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to to be used in routine histology and microbiology applications; and specifically in applications by delivering consistently crisp, featherededge blood smears with good morphology.
Have Questions?
Catalog number FKS12404070
Price (SGD)
13.00
PK
Add to cart
Price (SGD)
13.00
PK
Add to cart
  • Manufactured from Swiss extra-white soda-lime glass
  • Reduced iron content for low self-fluorescence
  • Greenish shimmer minimized
  • Guaranteed optimum, faultless transparency
  • Clean, flat and suitable for immediate use
Specifications
Edges90°, Ground
Length (Metric)76 mm
MaterialSoda Lime Glass
Thickness (Metric)1 mm
Width (Metric)26 mm
CoatingUncoated
Quantity50/Box, 2000 in outer carton OEM
Writing SurfaceNo
Dimensions (L x W)76 x 26 mm
Unit SizePK