Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Color |
---|---|---|
FKS12398208 | 250 mL | Amber |
FKS12388208 | 100 mL | Amber |
FKS12358208 | 100 mL | Clear |
FKS12368208 | 250 mL | Clear |
FKS12308218 | 500 mL | Amber |
FKS12378208 | 500 mL | Clear |