1x1 image pixel for data collection
Vacutainer™ Serum Tubes
Vacutainer™ Serum Tubes
Actual product may vary
Vacutainer™ Serum Tubes
Used to obtain a serum sample. BD™ Vacutainer™ Serum Tubes are made of durable PET plastic with a Hemogard™ closure. It has silica particles added to trigger coagulation, and are marked CAT and CE.
Have Questions?
Catalog NumberCapacity (Metric)Tube Size
FKS123969294 mL13 x 75 mm
Catalog number FKS12396929
Price (SGD)
90.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Capacity (Metric):
4 mL
Tube Size:
13 x 75 mm
Price (SGD)
90.00
PK
Add to cart
In order to obtain serum samples from plastic tubes, the tube must have a coagulation activator added. As the plastic surface alone is insufficient to trigger the coagulation within an acceptable time, BD Vacutainer™ plastic serum tubes have silica particles added for this purpose.
  • These tubes are marked with the acronym CAT (Clot Activator Tube)
  • Recommended minimum time for coagulation from patients who have not been treated with anticoagulants is 60 minutes
  • Centrifuging conditions: ≤1300 g for 10 minutes at 18–25°C
Specifications
Product TypeSerum Tube
AdditiveSilica
MaterialPET
Closure TypeHemogard
Capacity (Metric)4 mL
Closure ColorRed
Tube Size13 x 75 mm
For Use With (Application)Used to obtain a serum sample
Label TypeBlock
Certifications/ComplianceCE, CAT
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link