Check Order Status
Lids For Porcelain Crucibles
Actual product may vary
Fisherbrand
Lids For Porcelain Crucibles
Designed to fit Haldenwanger™ Porcelain Crucibles.
Have Questions?
Catalog Number
For Use With (Application)
FKS12396487
20mL crucibles
Catalog number
FKS12396487
Price (SGD)
306.00
PK
Add to cart
For Use With (Application):
20mL crucibles
Price (SGD)
306.00
PK
Add to cart
Available in a variety of sizes
For use with porcelain crucibles
Specifications
Color
White
Material
Porcelain
For Use With (Application)
20mL crucibles
Diameter (Metric) Outer
46mm
Product Type
Crucible Lids