Check Order Status
Squat Form Porcelain Crucibles Without Lid
Actual product may vary
Fisherbrand
Squat Form Porcelain Crucibles Without Lid
Use these dishes for a variety of applications. Haldenwanger™ Squat Form Porcelain Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.
Have Questions?
Catalog Number
Capacity (Metric)
FKS12396477
6 mL
Catalog number
FKS12396477
Price (SGD)
227.00
PK
Add to cart
Capacity (Metric):
6 mL
Price (SGD)
227.00
PK
Add to cart
Available in a variety of sizes
Sizes up to 175mL glazed except rim
1,000mL and larger are glazed except outside base
Complies with DIN VDE 0335, type C 100
Specifications
Material
Porcelain
Finish
Glazed
Capacity (Metric)
6 mL
Diameter (Metric) Outer
30mm
Height (Metric)
19mm
Product Type
Squat Form Porcelain Crucibles