Microscope Slides with Ground Edges
Microscope Slides with Ground Edges
Actual product may vary
Microscope Slides with Ground Edges
Wrapped in cellophane for improved cleanliness. These Microscope Slides with Ground Edges are washed and polished glass slides with 90° ground edge.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThickness (Metric)Frosted
FKS123931181.2 to 1.5 mmNot Frosted
FKS115622030.8 to 1 mmTwin Frosted
FKS123731180.8 to 1 mmNot Frosted
FKS123831181 to 1.2 mmNot Frosted
Catalog number FKS12393118
Price (SGD)
12.00
PK
Add to cart
Thickness (Metric):
1.2 to 1.5 mm
Frosted:
Not Frosted
Price (SGD)
12.00
PK
Add to cart
  • Glass 76 mm x 26 mm
  • Available in fully clear glass or frosted at one end
  • Ground on all four edges
Specifications
Edges90°, Ground
FrostedNot Frosted
Length (Metric)76 mm
MaterialGlass
PrecleanedYes
Thickness (Metric)1.2 to 1.5 mm
TypePlain Microscope Slide
Width (Metric)26 mm
ColorClear
Quantity50 Pk.
Unit SizePK