Semperguard™ Powder-Free Disposable Vinyl Gloves
Semperguard™ Powder-Free Disposable Vinyl Gloves
Actual product may vary
Fisherbrand
Semperguard™ Powder-Free Disposable Vinyl Gloves
Manufactured from a high quality vinyl formula, Semperguard™ Powder-Free Disposable Vinyl Gloves are tear-resistant, seamless gloves that are flat with a rolled edge.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FKS12392179Small
FKS12312189Large
FKS12302189Medium
FKS12201338X-Large
Catalog number FKS12392179
Price (SGD)
-
Size:
Small
Disposable transparent protective gloves are flat with a rolled edge.
  • Particularly skin-friendly for latex allergy sufferers - free of latex proteins and accelerators
  • Free of DOP (DEHP / diethylhexyl phthalate)
  • High quality vinyl (PVC) provides excellent wearing comfort
  • Excellent quality at an economical cost
  • CE category I Protective glove for minimal risks
  • Tested and certified according to VO 1935/2004 - suitable for contact with non-fatty foods
Specifications
Certifications/ComplianceCE Category I, EN 420, EN 374, EN 455
ClassificationCE category I Protective glove for minimal risks
ColorTransparent
DisposableSingle-use
Exterior FinishSmooth
HandAmbidextrous
Latex FreeYes
MaterialVinyl
PowderedPowder Free
SizeSmall
SterilityNon-sterile
Thickness (Metric)0.14 mm
Glove Cuff StyleRolled
Packaging TypeDispenser box
Palm TypeFlat
Length (Metric)240 mm
Unit SizePK