Lab Markers
Lab Markers
Actual product may vary
Lab Markers
Write on a variety of surfaces with these versatile lab markers. Cole-Parmer™ Lab Markers permanently mark plastic; wash off glass, metal, or porcelain. Markers won’t bleed or smudge when subjected to water or alcohol.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FKS12392028Green
FKS11700239Blue
FKS12372028Black
FKS11710239Red
Catalog number FKS12392028
Price (SGD)
143.00
PK
Add to cart
Color:
Green
Price (SGD)
143.00
PK
Add to cart
  • Fine-point fiber tip
  • Quick-drying ink
  • Writes on cold, wet surfaces
  • Available in four different colors
Specifications
ColorGreen
For Use With (Application)For Glass, Porcelain or Metal surfaces
Quantity10/Pk.
Unit SizePK