Check Order Status
Polyethylene Biohazard Bags
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Fisherbrand
Polyethylene Biohazard Bags
Use either plain or printed Alpha Packaging™ Polyethylene Biohazard Bags for specimen collection and transport. They are printed with a biohazard warning.
Have Questions?
Contact Us
Catalog Number
Width (Metric)
FKS12390529
150 mm
Catalog number
FKS12390529
Price (SGD)
35.00
PK
Add to cart
Width (Metric):
150 mm
Price (SGD)
35.00
PK
Add to cart
Plain or printed with a biohazard warning
Opening along width. 50μm thickness
Resealable section for the specimen
Specifications
Material
Polyethylene
Thickness (Metric)
50 μm
Width (Metric)
150 mm
Product Type
Biohazard Bag
Unit Size
PK
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link