Check Order Status
Modelling Clay
Actual product may vary
Fisherbrand
Modelling Clay
Looks and acts like
Plasticine
, a brand of modelling clay. Azpack™ Modelling Clay is stays moist and pliable.
Have Questions?
Catalog number
FKS12388049
Price (SGD)
-
Easily workable and pliable
Smooth texture
Odorless
Will not dry out
Color may stain fingers, but easily washes off
Non-toxic
CE marked
Conforms to EN71 safety regulations
Specifications
Packaging
500g bar
Quantity
1 Ea.
Product Type
Clay
Certifications/Compliance
CE Marked, EN71 safety
Color
Brown