Soda Lime Glass Reagent Bottles
Soda Lime Glass Reagent Bottles
Actual product may vary
Fisherbrand
Soda Lime Glass Reagent Bottles
Complies with ISO/EN recommendations. Poulten Graf™ Soda Lime Bottles have angled shoulders to ensure steady pouring and feature an interchangeable octagonal head.
Have Questions?
Catalog NumberCapacity (Metric)Color
FKS1238701830 mLClear
Catalog number FKS12387018
Price (SGD)
-
Capacity (Metric):
30 mL
Color:
Clear
  • Polypropylene stopper
  • Narrow neck construction
  • Specifications
    Capacity (Metric)30 mL
    ColorClear
    MaterialGlass
    MouthNarrow
    Product TypeReagent Bottle
    Closure TypeStopper
    Unit SizePK