Sharpsguard™ Orange Multi-Purpose Sharps Container with Pail Handle
Sharpsguard™ Orange Multi-Purpose Sharps Container with Pail Handle
Actual product may vary
Fisherbrand
Sharpsguard™ Orange Multi-Purpose Sharps Container with Pail Handle
Daniels Healthcare™ Sharpsguard™ Orange Multi-Purpose Sharps Containers are for disposal of sharps uncontaminated by medicinal products. Features include a robust pail handle for easy transportation and a wide aperture for easy insertion of larger items.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Length (Metric)Opening Size (Metric)
FKS1238243917.86 L343 mm327 mm58 mm x 148 mm (L x W)
FKS123724399.27 L249 mm275 mm48 mm x 148 mm (L x W)
Catalog number FKS12382439
Price (SGD)
33.00
EA
Add to cart
Capacity (Metric):
17.86 L
Height (Metric):
343 mm
Length (Metric):
327 mm
Opening Size (Metric):
58 mm x 148 mm (L x W)
Price (SGD)
33.00
EA
Add to cart
  • Container for disposal by incineration
  • Rigid polypropylene construction and wall thickness
  • Offers outstanding protection against penetrating sharps and leaking fluids
  • Wide aperture allows economical and convenient disposal of large items
  • When container is full to the fill line it must be locked and disposed of safely
  • All have clear British Standard markings, user instructions and a visible fill line
Specifications
Capacity (Metric)17.86 L
Certifications/ComplianceBS 7320: 1990, ISO 9002
Closure TypeLid
ColorYellow, Orange
Height (Metric)343 mm
Length (Metric)327 mm
Opening Size (Metric)58 mm x 148 mm (L x W)
AutoclavableAutoclavable
For Use With (Application)Sharps Container
MaterialPolypropylene
Product TypeSharps Container
Unit SizeEA