SuperFrost™ Microscope Slides, Ground 90°
Product Image
Fisherbrand
SuperFrost™ Microscope Slides, Ground 90°
The printed Epredia™ SuperFrost tab is impervious to virtually all common laboratory chemicals and reagents. A wide range of colors (7 standard colors) allows several examination specimens to be labeled and archived simply and clearly.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorQuantity
FKS12382098Blue50/Box, 2000 in outer carton
FKS12312108Green50/Box, 2000 in outer carton
Catalog number FKS12382098
Price (SGD)
-
Color:
Blue
Quantity:
50/Box, 2000 in outer carton
  • Time-consuming labeling with paper labels is no longer required
  • The writing area is resistant to all common solvents
  • SuperFrost slides may be sorted by color e.g. by patient, urgency, stain or examination method
  • Simple recognition of specified characteristics such as year of examination or department, use etc.
Specifications
Edges90°, Ground
Length (Metric)76 mm
Thickness (Metric)1 mm
Width (Metric)26 mm
ColorBlue
Certifications/ComplianceISO8037/I
CoatingUncoated
Quantity50/Box, 2000 in outer carton
Writing SurfaceYes
Dimensions (L x W)76 x 26 mm
Unit SizePK