Polyethylene, Aspirator Bottle
Polyethylene, Aspirator Bottle
Actual product may vary
Fisherbrand
Polyethylene, Aspirator Bottle
Capacity Metric: 5 L
Have Questions?
Catalog number FKS12380449
Price (SGD)
75.00
EA
Add to cart
Price (SGD)
75.00
EA
Add to cart
Capacity Metric: 5 L
  • Screw on cap
  • With carry handle
Specifications
Capacity (English)1.3 gal.
Capacity (Metric)5 L
Diameter (Metric) Outer170 mm
Height (Metric) With Closure332 mm
MaterialPolyethylene
Product TypeAspirator Bottle
Unit SizeEA