Plastipak™ Catheter-Tip Syringe
Plastipak™ Catheter-Tip Syringe
Actual product may vary
Fisherbrand
Plastipak™ Catheter-Tip Syringe
Graduated concentric catheter tip luer slip syringe. BD Biosciences™ Plastipak™ Catheter-Tip Syringe is sterile and single use.
Have Questions?
Catalog NumberVolume (Metric)
FKS12379149100 mL
Catalog number FKS12379149
Price (SGD)
626.00
PK
Add to cart
Volume (Metric):
100 mL
Price (SGD)
626.00
PK
Add to cart
  • 100ml
  • Sterile
  • Disposable
  • Graduated
  • Catheter tip
  • Luer slip
Specifications
Certifications/ComplianceCE Mark, European Directive 93/42 / EEC ( compliance procedure Annex V and VII) Certificates: No. 95 Q 252156 and No. 252231 Q (references 309661 & 309620 ) ; certified by NSAI (No. 0050)
MaterialPlastic
ColorTransparent
Diameter (Metric) Outer39 mm
Latex FreeYes
Length (Metric)122 mm
Product TypeCatheter-tip Syringe
DisposableSingle-use
Graduations1 mL
Volume (Metric)100 mL
Connection TypeCatheter