PIPETMANTM L Mechanical Pipet
PIPETMANTM L Mechanical Pipet
Actual product may vary
Fisher Catalogue
PIPETMANTM L Mechanical Pipet
The PIPETMANTM L Mechanical Pipet is a lightweight pipet with lockable volume and low forces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelVolume (Metric)AccuracyIncludes
FKS12376152P1000L100 to 1000 μL±3, ±4, ±8μLPlastic Tip Ejector
FKS12326152P100L10 to 100 μL±0.35, ±0.4, ±0.8μLStainless Steel Tip Ejector
FKS12346152P200L20 to 200 μL±0.5, ±0.8, ±1.6μLStainless Steel Tip Ejector
Catalog number FKS12376152
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Model:
P1000L
Volume (Metric):
100 to 1000 μL
Accuracy:
±3, ±4, ±8μL
Includes:
Plastic Tip Ejector
Mechanical pipette combines the legendary robustness of the PIPETMAN™ Classic with a newly designed body and state-of-the-art internal mechanism to offer the superior comfort and accuracy. Available in 8 single channel models covering the volume range of 0.2μL to 10mL.
  • Redesigned handle fits naturally in user's hand, no matter the size.
  • Volume locking system guarantees no accidental change in volume while pipetting.
  • Adjustable tip ejector provides comfort for right- and left-handed users.
  • 2D identification code allows easy tracking of product information.
  • New piston assembly designed to drastically improve pipetting and purge forces.
  • Redesigned tip ejector shape and internal mechanism offers reduced tip ejection forces by 50%.
Specifications
Accuracy±3, ±4, ±8μL
Compatible TipsD1000, D1200, DF1000ST, DF1200ST tip
Adjustable Tip SpacingNo
AutoclavableAutoclavable
ColorBlue
Dispensing SpeedManual
DisposableReusable
For Use WithD-1000, D-1200, DF1000ST, DF1200ST Tip
Grip TypeErgonomic
MaterialPolypropylene
ModelP1000L
Number of Channels1
Product LinePipetman
ProgrammabilityNo
SterilityNon-sterile
TypeMechanical Pipette
Volume (Metric)100 to 1000 μL
Finger StyleErgonomic
For Use With (Application)Transporting a measured volume of liquid
For Use With (Equipment)D1000, D1200, DF1000ST, DF1200ST tip
IncludesPlastic Tip Ejector
Quantity1
Warranty1 Year
Length (English)4.33 in.
Length (Metric)11 cm
Width (English)4.33 in.
Width (Metric)11 cm
Height (English)1.57 in.
Height (Metric)4 cm
Color CodeBlue
Fit for PurposeCertified Quality
Unit SizePK