Check Order Status
Quartz Wool
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Fisherbrand
Quartz Wool
Filter acids and hot gases or use as a stopper for combustion tubes. Azpack™ Quartz Wool can be exposed to temperatures up to 1050°C.
Have Questions?
Contact Us
Catalog number
FKS12373866
Price (SGD)
291.00
EA
Estimated availability date
01-Oct-2025
Add to cart
Price (SGD)
291.00
EA
Add to cart
A mass of fibres, average diameter 6μm.
Used for filtering acids and hot gases or as a plug for combustion tubes.
Specifications
Quantity
1Each
Product Type
Wool Filter
For Use With (Application)
Filtering acids, hot gases
Material
Quartz fiber
Weight (Metric)
10g
Melting Point
400°C
Diameter (Metric) Outer
6μm per fiber
Unit Size
EA
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link