1x1 image pixel for data collection
Hazardous Spill Response Kit
Hazardous Spill Response Kit
Actual product may vary
Fisherbrand
Hazardous Spill Response Kit
Contains chemical sorbents made from inert, synthetic fibers to be deployed wherever hazardous liquids are in use, for the protection of both employees and equipment. 3M™ Hazardous Spill Response Kits can be used to contain and clean up the hazardous liquid without delay.
Have Questions?
Catalog NumberCapacity (Metric)Dimensions (L x W)Includes
FKS123724595 L400 x 70 mm10 sheets, 1 pillow, 1 disposable bag with tie
Catalog number FKS12372459
Price (SGD)
-
Capacity (Metric):
5 L
Dimensions (L x W):
400 x 70 mm
Includes:
10 sheets, 1 pillow, 1 disposable bag with tie
Kits contain chemical sorbents made from inert, synthetic fibers to be deployed wherever hazardous liquids are in use, for the protection of both employees and equipment. In the event of a spill, the kit can be used to contain and clean up the hazardous liquid without delay.
  • Constructed from polypropylene fibers
  • Available in a wide range of formats
  • Lightweight and dust-free
  • High absorption capacity minimizes waste for disposal.
Specifications
Capacity (Metric)5 L
MaterialPolypropylene Fiber
TypeSpill Kit
Includes10 sheets, 1 pillow, 1 disposable bag with tie
Packaging Quantity1 Ea.
Dimensions (L x W)400 x 70 mm
Unit SizeEA
Share catalog number, name or link