A1B1E1K1 Gas Filter For 8000 Series Respiratory Cartridges
A1B1E1K1 Gas Filter For 8000 Series Respiratory Cartridges
Actual product may vary
Fisherbrand
A1B1E1K1 Gas Filter For 8000 Series Respiratory Cartridges
For protection against organic + inorganic gases and vapours
Have Questions?
Catalog NumberFilter Type
FKS12372309A1B1E1K1
Catalog number FKS12372309
Price (SGD)
251.00
PK
Add to cart
Filter Type:
A1B1E1K1
Price (SGD)
251.00
PK
Add to cart
Moldex 8000 Series ABEK1 gas filter (8900). ABEK1 gas filter for the 8000 Series half mask. For protection against organic + inorganic gases and vapours, boiling point >65°C, + ammonia and acid gases. 10 x WEL or 1,000ppm whichever is lower.
Specifications
Filter TypeA1B1E1K1
Product TypeGas Filter
For Use With (Equipment)For Series 8000 Respiratory Filter
Quantity10/Pk.
Unit SizePK