Check Order Status
Mercury Collector
Actual product may vary
Fisherbrand
Mercury Collector
Captures spilled mercury beads quickly and conveniently. Bel-Art™ Mercury Collector holds 125 ml of mercury.
Have Questions?
Change view
Catalog Number
For Use With (Application)
FKS12372088
Mercury spill clean-up replacement pad
FKS10595444
Mercury spill clean-up
2 Options
Catalog number
FKS12372088
Price (SGD)
167.00
PK
Add to cart
For Use With (Application):
Mercury spill clean-up replacement pad
Price (SGD)
167.00
PK
Add to cart
Captures spilled mercury beads quickly and conveniently. Bel-Art™ Mercury Collector holds 125 ml of mercury.
Lid features absorbent foam pad on interior surface
Replacement foam pad available
Dispose of mercury by approved disposal method
Specifications
Color
White
Capacity (Metric)
125 mL
Material
Polyethylene
Type
Replacement Pad
For Use With (Application)
Mercury spill clean-up replacement pad
Includes
Jar, perforated plate, lid with attached foam pad
Height (English)
1.88 in.
Height (Metric)
4.77 cm