Tube Brush
Tube Brush
Actual product may vary
Fisher Catalogue
Tube Brush
Features center set bristles. Pinnacle Brush™ Tube Brush is for cleaning tubes that are open at both ends.
Have Questions?
Catalog NumberDiameter (Metric) Bristle
FKS123683383 mm
Catalog number FKS12368338
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric) Bristle:
3 mm
  • Center set nylon bristles
  • Galvanized steel handle
Specifications
Product TypeTube Brush
Bristle MaterialNylon
Handle MaterialMetal
Diameter (Metric)0.3 cm
Diameter (Metric) Bristle3 mm
Length (Metric)44 cm
Length (Metric) Brush8 cm
Length (Metric) Handle19 cm, 18 cm
Quantity10 Pk.
SizeBrush Circumference: 1 cm
Width (Metric) Bristle3 mm
Unit SizePK