Check Order Status
Zinc Plated Stand Rods
Actual product may vary
Fisherbrand
Zinc Plated Stand Rods
An excellent option for the construction of retort stands in the laboratory. R&L Enterprises™ Zinc Plated Rods are manufactured from mild steel.
Have Questions?
Change view
Catalog Number
Length (Metric)
Diameter (Metric) Rod
FKS12368139
600 mm
12.5 mm
FKS12358139
500 mm
10 mm
2 Options
Catalog number
FKS12368139
Price (SGD)
157.00
PK
Add to cart
Length (Metric):
600 mm
Diameter (Metric) Rod:
12.5 mm
Price (SGD)
157.00
PK
Add to cart
Threaded
Available for a range of diameters and lengths
Length of rod does not include the thread
Specifications
Material
Zinc Plated Mild Steel
Length (Metric)
600 mm
Diameter (Metric) Rod
12.5 mm
Product Type
Rod