▸
Flasks
▸
Filtering Flasks
▸
Conical Filter Vacuum Flask with Hose Tubulature
Actual product may vary
Fisherbrand
Conical Filter Vacuum Flask with Hose Tubulature
Use this Conical Filter Vacuum Flask with Hose Tubulature with a vacuum funnel and filter. This thick-walled Ehlenmeyer flask features a glass tube protrusion about an inch from its neck and is also known as a Büchner flask.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FKS12363886
1000 mL
FKS12343886
250 mL
2 Options
Catalog number
FKS12363886
Price (SGD)
96.00
Each
Add to cart
Capacity (Metric):
1000 mL
Price (SGD)
96.00
Each
Add to cart
With 8mm outer diameter tubulature
Thick-walled borosilicate glass
Vacuum flask
Specifications
Diameter (Metric) Outer Tubing
8 mm
Capacity (Metric)
1000 mL
Material
Borosilicate Glass