Sharpsguard™ Polypropylene General Purpose Sharps Container
Sharpsguard™ Polypropylene General Purpose Sharps Container
Actual product may vary
Fisherbrand
Sharpsguard™ Polypropylene General Purpose Sharps Container
Use this general-purpose sharps container for outstanding protection against penetrating sharps and leaking fluids. Daniels Healthcarer™ Sharpsguard™ Polypropylene General Purpose Sharps Containers feature rigid polypropylene construction and wall thickness designed to meet demands for strength, simplicity and value.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Length (Metric)Certifications/Compliance
FKS123624397 L205 mm248 mmBS 7320: 1990, ISO 9002
FKS123324392.5 L163 mm176 mmBS 7320: 1990, ISO 9002
FKS123424393.75 L218 mm176 mmBS 7320: 1990, ISO 9002
FKS123524395 L285 mm176 mmBS 7320: 1990, ISO 9002
Catalog number FKS12362439
Price (SGD)
12.00
EA
Add to cart
Capacity (Metric):
7 L
Height (Metric):
205 mm
Length (Metric):
248 mm
Certifications/Compliance:
BS 7320: 1990, ISO 9002
Price (SGD)
12.00
EA
Add to cart
  • Maximum capacity in the minimum working space
  • Manufactured to BS 7320: 1990, ISO 9002 standards and UN Packing Group II approved
  • All have clear British Standard markings, user instructions and a visible fill line
  • A robust side handle prevents the container rolling for safe transportation and ease of disposal
  • Handle prevents the container rolling the unlikely event it is dropped, and allows the container to be hung onto a POUDS™ bracket
Specifications
Capacity (Metric)7 L
Certifications/ComplianceBS 7320: 1990, ISO 9002
Closure TypeLid
ColorYellow, Red
Height (Metric)205 mm
Length (Metric)248 mm
Opening Size (Metric)77 x 45 mm (L x W)
AutoclavableAutoclavable
For Use With (Application)For easy transportation