Frosted Microscope Slides, Ground 90°
Product Image
Fisherbrand
Frosted Microscope Slides, Ground 90°
Premium slides are manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to to be used in routine histology and microbiology applications; and specifically in applications by delivering consistently crisp, featherededge blood smears with good morphology.
Have Questions?
Catalog NumberThickness (Metric)FrostedQuantityWriting Surface
FKS123620981 to 1.2 mm20 mm50/Box, 2000 in outer cartonYes
Catalog number FKS12362098
Price (SGD)
15.00
PK
Add to cart
Thickness (Metric):
1 to 1.2 mm
Frosted:
20 mm
Quantity:
50/Box, 2000 in outer carton
Writing Surface:
Yes
Price (SGD)
15.00
PK
Add to cart
  • Manufactured from Swiss extra-white soda-lime glass
  • Reduced iron content for low self-fluorescence
  • Greenish shimmer minimized
  • Guaranteed optimum, faultless transparency
  • Clean, flat and suitable for immediate use
Specifications
Frosted20 mm
Length (Metric)76 mm
MaterialSoda Lime Glass
Thickness (Metric)1 to 1.2 mm
Width (Metric)26 mm
CoatingUncoated
Quantity50/Box, 2000 in outer carton
Writing SurfaceYes
Dimensions (L x W)76 x 26 mm
Unit SizePK