Porcelain Evaporating Basins
Porcelain Evaporating Basins
Actual product may vary
Fisherbrand
Porcelain Evaporating Basins
Glazed inside and out
Have Questions?
Catalog NumberCapacity (Metric)
FKS1236070970 mL
Catalog number FKS12360709
Price (SGD)
84.00
EA
Add to cart
Capacity (Metric):
70 mL
Price (SGD)
84.00
EA
Add to cart
Deep form, French shape, glazed inside and out.
Specifications
Bottom ShapeRound
Capacity (Metric)70 mL
MaterialPorcelain
ShapeRound
TypeEvaporating Basin
Interior FinishGlazed
Height (Metric)40 mm
Diameter (Metric)75 mm
Unit SizeEA