Tough Spots™ Tube Labels
Tough Spots™ Tube Labels
Actual product may vary
Fisherbrand
Tough Spots™ Tube Labels
Ensure microtube labels stay put through any lab procedure. Tough Spots™ Tube Labels won’t peel or migrate, even with exposure to extreme temperatures, caustic agents or centrifugation. Pre-cut round labels in two sizes are available in a variety of colors. 1,000 labels per roll.
Have Questions?
Catalog NumberColorDiameter (Metric)
FKS12358817White11 mm
Catalog number FKS12358817
Price (SGD)
198.00
EA
Add to cart
Color:
White
Diameter (Metric):
11 mm
Price (SGD)
198.00
EA
Add to cart
Superior to conventional tapes and labels for use in research and clinical laboratories.
  • Constructed of thin, chemically inert polyolefin with strong acrylic adhesive.
  • Adheres to most plastic, glass and metal mictrotubes and other items.
  • Labels won't peel or migrate so they can slide easily in centrifuge rotors without binding or catching.
  • Wide assortment of colors for easy sample or researcher identification.
  • Round, pre-cut, peel-off labels for tube tops in convenient dispensing rolls.
  • Sized for microcentrifuge tubes.
    • 9.5 mm diameter Tough Spots are ideal for 0.5 mL microtubes.
    • 13 mm diameter Tough Spots are ideal for 1.5 / 2.0 mL tubes.
Withstands extreme environments so tubes stay clearly identified.
  • Heat: steam autoclaving, boiling water
  • Cold: freezer temperatures to -80°C
  • Organic solvents and caustic agents
  • Centrifugation
Specifications
AutoclavableAutoclavable
Adhesive TypeAcrylic
ColorWhite
DisplayBlank
FinishMatte
FormatRoll
MaterialPolyolefin
Printable SidesSingle-side Printable
Product TypeLabels
SterilityNon-sterile
For Use With (Application)Microcentrifuge tubes and other containers in the research and clinical laboratory
For Use With (Equipment)0.5 mL Tubes
Labels per Row2
Quantity1000/Pk.
ShapeRound
Temperature Range (Metric)-196°C to +80°C
Dimensions (LxW)1 x 1 cm
Length (English)0.4 in.
Width (English)0.44 in.
Width (Metric)11 mm
Diameter (English)0.44 in.
Diameter (Metric)11 mm
Unit SizeEA