Clear Glass Extra Wide Neck Bottles with Polypropylene Screw Caps
Clear Glass Extra Wide Neck Bottles with Polypropylene Screw Caps
Actual product may vary
Fisher Catalogue
Clear Glass Extra Wide Neck Bottles with Polypropylene Screw Caps
Write on the white caps with marker for labeling purposes. Azpack™ Clear Glass Extra Wide Neck Bottles with Polypropylene Screw Caps are manufactured from soda lime glass.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FKS1235716830 mL38/R3
FKS1135097560 mL51/R3
FKS11398033120 mL58/R3
Catalog number FKS12357168
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
30 mL
Closure Size:
38/R3
  • Soda lime glass bottle
  • Extra wide neck
  • Includes unlined polypropylene cap
  • White cap is ideal for use with marker pen
Specifications
Capacity (Metric)30 mL
ColorClear
MaterialSoda Lime Glass
Quantity20/PK
Closure ColorWhite
Closure IncludedYes
MouthExtra Wide
Height (Metric)48 mm
Closure MaterialPolypropylene
Closure Size38/R3
Diameter (Metric)41 mm
Unit SizePK