Use these boats for a variety laboratory applications. Haldenwanger™ Aluminum Oxide Combustion Boats are ideal for chemical calcining, metal casting and sample preparation.
Have Questions?
Change view
Catalog Number
Dimensions (L x W x H)
FKS12357018
160 x 40 x 21 mm
FKS12347018
120 x 30 x 15 mm
2 Options
Catalog number FKS12357018
Price (SGD)
606.00
EA
Add to cart
Dimensions (L x W x H):
160 x 40 x 21 mm
Price (SGD)
606.00
EA
Add to cart
Aluminum oxide boats
Great hardness and strength
Resistant to heat and caustic chemicals
Suitable for continuous use up to 1,750°C
Specifications
TypeCombustion Boat
Dimensions (L x W x H)160 x 40 x 21 mm
For Use With (Application)For continuous use up to 1700 to 1750°C.