Check Order Status
Cork Rings
Actual product may vary
Fisherbrand
Cork Rings
Ring to support flasks
Have Questions?
Change view
Catalog Number
Diameter (Metric) Inner
Diameter (Metric) Outer
FKS12356447
85 mm
115 mm
FKS12346447
45 mm
75 mm
2 Options
Catalog number
FKS12356447
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric) Inner:
85 mm
Diameter (Metric) Outer:
115 mm
Ring to support flasks
Available in different diameters
Specifications
Product Type
Cork Ring
For Use With (Application)
Flask Support
Quantity
1 Each
Depth (Metric)
38 mm
Diameter (Metric) Outer
115 mm
Diameter (Metric) Inner
85 mm