Check Order Status
Disposable Hemacytometers, Neubauer Improved Grid
Actual product may vary
Fisherbrand
Disposable Hemacytometers, Neubauer Improved Grid
These Disposable Hemacytometers with Neubauer Improved Grid feature a precise fixed-depth counting chamber with superior accuracy and reproducibility.
Have Questions?
Catalog Number
Height (Metric)
FKS12353800
100 μm
Catalog number
FKS12353800
Price (SGD)
1,630.00
PK
Add to cart
Height (Metric):
100 μm
Price (SGD)
1,630.00
PK
Add to cart
Cole-Parmer Durable Plastic Hemacytometer, Height: 100um, Disposable: Yes, Quantity: 50Pack
Single use.
Light and unbreakable.
No coverslips required.
Disposable hemocytometer is made of durable plastics used for manual cell counting.
It consists of surface-patterned two enclosed 100 μm height chambers with two ports for sample injection.
It is a significant improvement of the conventional glass hemocytometer.
Single usage reduces the risk of exposure to potential infectious material, increases productivity by eliminating cleaning and work interruptions.
Specifications
Product Type
Plastic Hemacytometer
Grid Type
Neubauer
Disposable
Single-use
Material
Durable Plastic
Quantity
50 Pk.
Height (English)
0.004 in.
Height (Metric)
100 μm
For Use With (Application)
Cell Counting