Check Order Status
20mL Borosilicate Glass Scintillation Vials with White Urea Caps
Actual product may vary
Fisherbrand
20mL Borosilicate Glass Scintillation Vials with White Urea Caps
Fit all common counters
Have Questions?
Change view
Catalog Number
Material (Liner)
Closure Type
FKS12353317
Foil/Cork
Attached Screw Cap
FIS03-337-7
Polyethylene
Separate Screw Cap
FIS03-337-5
Foil/Cork
Attached Screw Cap
3 Options
Catalog number
FKS12353317
Price (SGD)
856.00
EA
Add to cart
Material (Liner):
Foil/Cork
Closure Type:
Attached Screw Cap
Price (SGD)
856.00
EA
Add to cart
Fit all common counters
Potassium-free glass provides consistently low activity counts and excellent light transmission
Caps fit GPI 22-400 thread finish and are suitable for marking
Specifications
Material (Liner)
Foil/Cork
Diameter (English) Outer
1.12 in.
Color
White
Diameter (Metric) Outer
28 mm
Material
Borosilicate Glass
Volume (Metric)
20 mL
Height (English)
2.37 in.
Packaging Type
Shrink-wrapped Tray
Volume (English)
0.67 oz.
Volume (Metric) Total
20 mL
Closure Material
Urea
Closure Type
Attached Screw Cap
Height (Metric)
61 mm
Type
Scintillation Vial
Closure Size
22-400
Marking Spot
Yes