20mL HDPE Scintillation Vials with Polypropylene Cap
20mL HDPE Scintillation Vials with Polypropylene Cap
Actual product may vary
Fisherbrand
20mL HDPE Scintillation Vials with Polypropylene Cap
Fit all common counters
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging Type
FKS12351599Bulk, Caps attached
FIS03-337-23ABulk, Caps attached
Catalog number FKS12351599
Price (SGD)
508.00
EA
Add to cart
Packaging Type:
Bulk, Caps attached
Price (SGD)
508.00
EA
Add to cart
Fit all common counters
White polypropylene caps.
  • Provide low background counts and excellent light transmission
  • Resist leaking and swelling caused by solvent degradation
  • Caps fit GPI 22-400 thread, and are suitable for marking
Specifications
Material (Liner)Metal Foil/Pulp
Diameter (English) Outer1.1 in.
ColorWhite
Diameter (Metric) Outer28 mm
MaterialHDPE
Volume (Metric)20 mL
For Use With (Application)Provide low background counts and excellent light transmission, fit all common counters
Height (English)2.4 in.
Packaging TypeBulk, Caps attached
Volume (English)0.67 oz.
Volume (Metric) Total20 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap
Height (Metric)61 mm
TypeScintillation Vial
Closure Size22-400
Marking SpotYes
Unit SizeEA