Check Order Status
Azlon™ Ampoule Breaker
Actual product may vary
Fisherbrand
Azlon™ Ampoule Breaker
Break glass ampoules safely with easy and safe disposable of the broken glass . The Azlon™ ampoule breaker comes in a choice of sizes.
Have Questions?
Catalog Number
For Use With
FKS12351168
5-10 mL Ampoule
Catalog number
FKS12351168
Price (SGD)
1,985.00
EA
Add to cart
For Use With:
5-10 mL Ampoule
Price (SGD)
1,985.00
EA
Add to cart
Enables safe and easy breaking of glass ampoules
Broken ampoule top is retained for safe disposal
Choice of size to suit all common glass ampoules
Specifications
Color
Natural
Product Type
Ampule Breaker
Material
HDPE
For Use With
5-10 mL Ampoule