Wood Test Tube Holders
Wood Test Tube Holders
Actual product may vary
Fisherbrand
Wood Test Tube Holders
Use these test tube holders for general laboratory applications. S Murray™ Wood Test Tube Holders are designed for use with tubes up to 19mm in diameter.
Have Questions?
Catalog NumberPositions
FKS123493491
Catalog number FKS12349349
Price (SGD)
40.00
PK
Add to cart
Positions:
1
Price (SGD)
40.00
PK
Add to cart
Use these test tube holders for general laboratory applications. S Murray™ Wood Test Tube Holders are designed for use with tubes up to 19mm in diameter.
  • Wood with steel spring
  • Pack of 10
Specifications
Product TypeTube Holder
HoldsTubes (19 mm)
MaterialWood with Steel Spring
Positions1
For Use With (Equipment)19mm Tube
Quantity10 Pack
Diameter (Metric) Opening19 mm
Unit SizePK