Check Order Status
Stainless Steel Surgical Scalpel Handle 4GS
Actual product may vary
Fisherbrand
Stainless Steel Surgical Scalpel Handle 4GS
Individually wrapped. Swann Morton Stainless Steel Surgical Scapel Handles contain graduations on one side to assist with training, marking of surgical site or trauma injury investigation.
Have Questions?
Catalog Number
Size
FKS12348019
No. 4
Catalog number
FKS12348019
Price (SGD)
14.00
EA
Add to cart
Size:
No. 4
Price (SGD)
14.00
EA
Add to cart
Stainless Steel
Individually wrapped
Specifications
Size
No. 4