Universal pH Indicator Paper
Universal pH Indicator Paper
Actual product may vary
Fisherbrand
Universal pH Indicator Paper
Gives rough knowledge of pH.
Have Questions?
Catalog NumberpH RangeFormatLength (Metric)Quantity
FKS123453781 to 11Tube50 mm10 Pack
Catalog number FKS12345378
Price (SGD)
26.00
PK
Add to cart
pH Range:
1 to 11
Format:
Tube
Length (Metric):
50 mm
Quantity:
10 Pack
Price (SGD)
26.00
PK
Add to cart
  • Provides rapid and precise readings
  • Includes color scale printed on cover for convenience
Applications:
  • Used for general analysis in chemical, school laboratories
Specifications
pH Range1 to 11
FormatTube
Quantity10 Pack
TypeGrade 911
Length (Metric)50 mm
Width (Metric)10 mm
Unit SizePK