Spreader Blue Polystyrene Wedge-Shaped Spreader
Spreader Blue Polystyrene Wedge-Shaped Spreader
Actual product may vary
Fisherbrand
Spreader Blue Polystyrene Wedge-Shaped Spreader
High impact wedge-shaped disposable bacterial spreaders
Have Questions?
Catalog NumberQuantity
FKS12342048100 bags of 5
Catalog number FKS12342048
Price (SGD)
168.00
PK
Add to cart
Quantity:
100 bags of 5
Price (SGD)
168.00
PK
Add to cart
  • High impact polystyrene disposable bacterial spreaders.
  • Wedge shaped to reduce tendency to flip, or traditional “L” shape.
  • Packs of 500 contain re-sealable inner bags of either 20 or 5 sterile spreaders.
  • Manufactured under aseptic conditions
Specifications
ColorBlue
DisposableSingle-use
Quantity100 bags of 5
ShapeWedge-shaped
MaterialPolystyrene
Product TypeSpreader
For Use With (Application)High impact Polystyrene disposable bacterial spreaders
Unit SizePK