Suitable for sealing laboratory containers such as flasks, beakers, bottles, tubes and others. Bryson Packaging™ Cling Film Rolls are supplied in boxes with a cutter edge.
Have Questions?
Catalog Number
Width (Metric)
FKS12338039
300mm
Catalog number FKS12338039
Price (SGD)
88.00
EA
Add to cart
Width (Metric):
300mm
Price (SGD)
88.00
EA
Add to cart
Made from extruded PVC film, 50μm thick
Suitable for use in deep freezers
Two-way stretch
Staphylococci-proof
Heat destruction at 125°C
Specifications
ApplicationsSuitable for Sealing laboratory Containers - Flasks, Beakers, Bottles, Tubes and for use in deep freeze