Screw Caps for Disposable Glass Tubes
Screw Caps for Disposable Glass Tubes
Actual product may vary
Fisherbrand
Screw Caps for Disposable Glass Tubes
Phenolic caps have cemented-in rubber liners
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorClosure SizeMaterial
FKS12337279Black15-415Phenolic
FIS14-957-82ABlack13-415Phenolic
FIS14-959-36ABlack15-415Phenolic
Catalog number FKS12337279
Price (SGD)
-
Color:
Black
Closure Size:
15-415
Material:
Phenolic
Phenolic caps have cemented-in rubber liners
  • Autoclavable
  • Polypropylene cap is linerless
Specifications
Closure Size15-415
Liner MaterialRubber
Pre-sterilizedNo
Product TypeScrew Cap
AutoclavableAutoclavable
MaterialPhenolic
ColorBlack
Tip MaterialRubber
SterilityNon-sterile
For Use With (Equipment)14-959-35A, 14-959-35B, 14-959-35AA
Unit SizeEA