uPIPETTE Adjustable Single Channel Pipette
uPIPETTE Adjustable Single Channel Pipette
Actual product may vary
Fisher Catalogue
uPIPETTE Adjustable Single Channel Pipette
Ergonomic design reduces fatigue and your risk of repetitive stress injuries while providing superior accuracy and precision.
Have Questions?
Catalog NumberVolume (Metric)
FKS1233701820 to 300 μL
Catalog number FKS12337018
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
20 to 300 μL
  • Compatible with Biotix uTIP™ and other universal pipette tips
  • Superior accuracy and precision with better ergonomics
  • Low drag seals and light spring forces produce a smooth draw and increased control
  • Easy to reach ejector button is cushioned to reduce thumb impact and stress when ejecting tips
  • Volume lock ensures accurate and precise measurements without volume drift
  • Superior accuracy and precision for more reliable data
  • Specifications
    Certifications/ComplianceISO 8655
    AutoclavableAutoclavable
    ColorGray
    DisposableReusable
    For Use WithuTIP Pipette Tip
    Grip TypeErgonomic
    MaterialPVDF
    ModelBiotix uPIPETTE
    Number of Channels1
    ProgrammabilityNo
    SterilityNon-sterile
    TypeAdjustable Single Channel Pipette
    Volume (Metric)20 to 300 μL
    IncludesCalibration Certificate
    Length (Metric)6 cm
    Width (Metric)20 mm
    Unit SizeEA