Nitrilite™ 93-311 Series Natural Nitrile Gloves
Nitrilite™ 93-311 Series Natural Nitrile Gloves
Actual product may vary
Fisherbrand
Nitrilite™ 93-311 Series Natural Nitrile Gloves
Produced in a controlled powder-free environment, featuring exceptionally low levels of extractable and particles.
Have Questions?
Catalog NumberSize
FKS12336347Small
Catalog number FKS12336347
Price (SGD)
-
Size:
Small
  • 100% nitrile polymer prevents risk of allergies related to latex protein
  • Nitrile coating
  • Textured fingertips grip design
  • Rolled beaded cuff
Specifications
ClassificationPPE Category II
ColorNatural
DisposableSingle-use
Exterior FinishSmooth
HandAmbidextrous
MaterialNitrile
PowderedPowdered
Product TypeControlled Environment Gloves
SizeSmall
SterilityNon-sterile
Thickness (Metric)0.12 mm
Glove Cuff StyleRolled
Quantity100 Pk.
Resistant ToAcids, alcohols, ketones and alkalis
Length (Metric)24 cm
Unit SizePK