AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves
AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves
Actual product may vary
AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves
AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves
AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves
AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves
Fisherbrand
AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves
Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.
Have Questions?
Catalog NumberSize
FKS123336099
Catalog number FKS12333609
Price (SGD)
-
Size:
9
Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.
  • Provide flexibility and tactility
  • Made from an exceptionally thin chemical-resistant, five-layer laminated film
  • Integral, nonwoven liner helps absorb and dissipate perspiration for a cooler, more comfortable fit
  • Can be worn as liners under heavier nitrile, neoprene, or PVC gloves
  • Hand specific
  • Broad spectrum chemical resistance
  • Wing-thumb for added wearer comfort
Specifications
ANSI Abrasion RatingANSI 0
Certifications/Compliance (Europe)CE 0493, EN420:2003 + A1:2009; Category lll
ApplicationsIndustrial
ColorWhite
HandSpecific
Latex FreeYes
MaterialLaminated Film
Product TypeChemical-resistant Gloves
Size9
Thickness (English)2.5 mil
Thickness (Metric)0.06 mm
EN388 StandardEN 388:0000
EN 421No
EN ISO 374-1Type A
EN ISO 374-1 CHEMICAL LETTER CODESABCEFGHIMS
EN ISO 374 - 5Bacteria, Fungi WITH Virus Protection
Glove Cuff StyleGauntlet
Grip DesignSmooth
LiningUnlined
Model2-100 9 902212
SeriesAlphaTec™ 02-100
Length (English)15 in.
Length (Metric)38 cm
Unit SizeEA