Check Order Status
Checker™ pH Tester and Replacement Part
Actual product may vary
Fisherbrand
Checker™ pH Tester and Replacement Part
For use with virtually any electrode that shares same connector.
Have Questions?
Catalog Number
For Use With (Application)
FKS12333408
pH
Catalog number
FKS12333408
Price (SGD)
205.00
EA
Add to cart
For Use With (Application):
pH
Price (SGD)
205.00
EA
Add to cart
pH range from 0.00 to 14.00
Provides high accuracy with 0.1 pH resolution
1.5V
RH max 95%
With LCD and manual two point calibration
3000 hours of battery life
Specifications
For Use With (Application)
pH
Accuracy
±0.2pH
Detection Range
0 to 14 pH