Glass Circle Coverslips
Glass Circle Coverslips
Actual product may vary
Fisher Catalogue
Glass Circle Coverslips
Glass Circle Coverslips are available in two thicknesses and a variety of diameters. Packed in plastic boxes to prevent breakage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThickness (Metric)Diameter (Metric)
FKS12333138#1 (0.13 to 0.16 mm)22 mm
FKS12323138#1 (0.13 to 0.16 mm)19 mm
Catalog number FKS12333138
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Thickness (Metric):
#1 (0.13 to 0.16 mm)
Diameter (Metric):
22 mm
These Glass Circle Coverslips are available in two thicknesses and a variety of diameters. Packed in plastic boxes to prevent breakage.
  • Manufactured from clear soda glass
  • Consistent in cut, thickness and dimensional accuracy
  • Free from blisters and flaws
Specifications
Product TypeCoverslip
ShapeCircle
MaterialSoda Lime Glass
Thickness (Metric)#1 (0.13 to 0.16 mm)
Diameter (Metric)22 mm
Quantity200/Pk.
Unit SizeEA