1x1 image pixel for data collection
Standard Chemical Sorbents
Standard Chemical Sorbents
Actual product may vary
Standard Chemical Sorbents
Standard Chemical Sorbents
Fisherbrand
Standard Chemical Sorbents
Absorb virtually any liquid with these chemical sorbents
Have Questions?
Catalog NumberCapacity (English)TypeModel
FKS123324598 gal.Chemical Sorbent PillowP300
Catalog number FKS12332459
Price (SGD)
1,270.00
PK
Add to cart
Capacity (English):
8 gal.
Type:
Chemical Sorbent Pillow
Model:
P300
Price (SGD)
1,270.00
PK
Add to cart
Meltblown polypropylene microfibers absorb liquids such as hydrofluoric acid and other aqueous-based chemicals, and soak up an average of 10 times their weight of most EPA-designated hazardous liquids.
  • Sorbents are dirt- and dust-free; contain no abrasive granules that can damage expensive equipment
  • Polypropylene base is chemically inert and incinerable; allows separation and recycling in many applications
  • Easy-to-spot yellow cartons protect sorbents from temperature and humidity for secure, long-term storage
  • Convenient dispenser boxes—suitable for tabletops or wall mounting—keep pads and pillows within easy reach Pads or rolls are ideal for use on work surfaces and can be trimmed to size
  • Mini booms surround and contain spills; pillows soak them up
  • 3M Catalog# P110, P200, P208, P300, P500
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
Certifications/ComplianceOSHA 29 CFR 1910.132
ColorYellow
Capacity (English)8 gal.
Capacity (Metric)30.3 L
MaterialPolypropylene
TypeChemical Sorbent Pillow
For Use With (Application)For control of spills of certain hazardous liquids
ModelP300
Dimensions (L x W)38 x 17.78 cm (15 x 7 in.)
Length (English)15 in.
Length (Metric)38 cm
Width (English)7 in.
Width (Metric)17.78 cm
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link