Vibrasorb™ Premium Vibration Damping Mounts
Actual product may vary
Vibrasorb™ Premium Vibration Damping Mounts
Vibrasorb™ Premium Vibration Damping Mounts
Fisherbrand
Vibrasorb™ Premium Vibration Damping Mounts
Protects electromechanical and optical equipment from exposure to vibration. Premium vibration damping mounts facilitates optimal equipment use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWidth (English)
FKS1233067935 x 35 x 7.6 cm
FKS1232067945 x 56 x 7.6 cm
Catalog number FKS12330679
Price (SGD)
6,408.00
Each
Width (English):
35 x 35 x 7.6 cm
Price (SGD)
6,408.00
Each
Protects electromechanical and optical equipment from exposure to vibration. Bel Art™ SP Scienceware™ Vibrasorb™ Premium Vibration Damping Mounts facilitates optimal equipment use.
  • Polished surface resists scratches and chemical attack 6.4cm (2½ in.) thick platforms
  • Molded of dense black & white terrazzo
  • Four Vibro-Absorber™ cushions with neoprene feet support up to 16kg (35 lb)
  • Larger size will absorb vibrations down to 13 Hz with a 16kg (35lb) load; smaller size will absorb vibrations down to 16 Hz. With a 16kg (35lb) load
  • With lighter loads, the minimum frequency of damping for both sizes will increase to a value between 16 Hz. And 25 Hz
  • Shipping weights: large 39kg (87lb), small 19kg (42lb)
Specifications
AutoclavableNot Autoclavable
Width (English)35 x 35 x 7.6 cm
Width (Metric)35 cm
Product TypeVibration Damping Mount
ColorGray
MaterialTerrazo
Unit SizeEach