Clean and disinfect work surfaces and equipment in a single, one-step operation. Day Impex™ Virkon™ Disinfectant Virucidal Tablets provide the widest proven spectrum of any disinfectant.
Have Questions?
Change view
Catalog Number
Quantity
FKS12328667
10 Tablets/Tub, 20 Tubs/Cs.
FKS12318667
50 Tablets/Tub, 12 Tubs/Cs.
2 Options
Catalog number FKS12328667
Price (SGD)
637.00
EA
Add to cart
Quantity:
10 Tablets/Tub, 20 Tubs/Cs.
Price (SGD)
637.00
EA
Add to cart
Use biocides safely. Always read the label and product information before use.
Widest proven spectrum of any disinfectant - proven effective in independent tests against all 18 virus families affecting man and animals, including AIDS (HIV) and Hepatitis B
Excellent bactericide and fungicide - cleans and disinfects work surfaces and equipment in a single, one-step operation
Can be applied in powder form direct to spillages of blood and other body fluids
Easily dissolves to make a liquid disinfectant
Odourless, non-irritant to skin and eyes, non-toxic vapour phase, exceptionally low oral toxicity
Under COSHH requirements, no maximum exposure limit is required
Non-corrosive, non-bleaching and biodegradable
Any viral disease transmitted via surfaces or instruments contaminated with infected blood, body fluids, faeces or saliva can be controlled by correct application For cleaning and disinfecting work surfaces and equipment.
Specifications
Product TypeVirucidal Disinfectant
FormTablet
For Use With (Application)For Cleaning and Disinfecting Work Surfaces and Equipment