Search Thermo Fisher Scientific
Catalog Number | Length (Metric) | Diameter (Metric) Inner | Description |
---|---|---|---|
FKS12328149 | 600 mm | 12.5 mm | Threaded |
FKS11785386 | 500 mm | 10 mm | Threaded |
FKS11759327 | 500 mm | 12.5 mm | Plain (without thread) |
FKS11705396 | 1000 mm | 12.5 mm | Threaded |