B-safe™ Polyethylene Biohazard Bag
B-safe™ Polyethylene Biohazard Bag
Actual product may vary
Fisherbrand
B-safe™ Polyethylene Biohazard Bag
Manufactured from polyethylene. Alpha Packaging™ B-safe™ Polyethylene Biohazard Bags effectively contain biohazard waste.
Have Questions?
Catalog NumberHeight (Metric)
FKS123206091015 mm
Catalog number FKS12320609
Price (SGD)
258.00
PK
Add to cart
Height (Metric):
1015 mm
Price (SGD)
258.00
PK
Add to cart
Manufactured from polyethylene. Alpha Packaging™ B-safe™ Polyethylene Biohazard Bags effectively contain biohazard waste.
  • Comes in loose packs or benchtop dispenser boxes.
  • For heavy-duty disposal
  • Marked on both sides with biohazard symbol and warnings:
    • "Biohazard, Danger of Infection"
    • "Burn Without Opening"
  • Warning in English, French, and German
  • Polyethylene (500 gauge)
Specifications
ColorYellow
DisposableSingle-use
Height (Metric)1015 mm
Length (Metric)790 mm
Length (English)31 in.
Height (English)39.9 in.
MaterialPolyethylene
Product TypeBiohazard Bag
Unit SizePK